Learn More
Fisherbrand™ Termómetros sin mercurio Ertco™ Exact-Temp™
Descripción
High-precision instrument to monitor temperature chambers, mounted in a shatterproof plastic bottle filled with liquid to ensure accurate readings.
- Traceable to international standards (with certificate of conformance and accuracy)
- Conventional thermometer works independent of any mains or battery connection
- For accurate measurements, even with opening and closing of the appliance door
- With engraved stems and thoroughly annealed for accuracy
- Organic, mercury-free filling colored red for easy readings
- Coated with PTFE resin to absorb shock and to contain glass and liquid in case of damage
- With individual serial numbers
- With double magnet for wall or door mounting
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™
Especificaciones
Especificaciones
| Relleno del frasco | Ethylene Glycol |
| Tamaño del frasco | 30 mL |
| Para utilizar con (aplicación) | Temperature Monitoring |
| Longitud total (imperial) | 4.92 in. |
| Longitud total (métrico) | 130 mm |
| Material | Glass, PETG |
| Relleno del termómetro | Líquido orgánico rojo |
| Tipo de producto | Bottle Thermometer |
| Temperatura máxima | 0 °C |
| Temperatura mín. (métrico) | -30°C |
| Mostrar más |