Learn More
EVX2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Novus Biologicals NBP1-90973
 Ver más versiones de este producto
Código de producto. 18720854
  • ONLINE EXCLUSIVE PRICE
    624.00€
    590.10€ / 0.10 ml
    Ahorro 33.90€
    5% Descuento
Descripción
EVX2 Polyclonal specifically detects EVX2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Immunohistochemistry, Immunohistochemistry (Paraffin)
RUO
IgG
This antibody was developed against Recombinant Protein corresponding to amino acids:PLHSALGELPAKGKFEIDTLFNLQHTGSESTVSSEISSAAESRKKPGHYS
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones