Learn More
Heidolph™ Matraz de evaporación
Código de producto. 10186781 Tienda Heidolph Instruments Productos
Click to view available options
Volumen (métrico):
100 mL
250 mL
500 mL
1000 mL
Unit Size:
1 PCS
cada uno
This item is not returnable. View return policy
Product Code. 10186781
Brand: Heidolph™ 5147200000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
For use with rotary evaporators
Los matraces de evaporación se suministran con un tamaño de junta NS 29/32 estándar, pero también están disponibles en NS 24/29
TRUSTED_SUSTAINABILITY
Specifications
Material Vidrio de borosilicato
Tipo Matraz de evaporación
Volumen (métrico) 250 mL
Tensión 115/230 VCA
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.