missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: YLGEHSSVFQ QPLDICHSFT SQGGGREPLP APYQHQLSEP CPPYPQQSFK QEYHDPLYEQ AGQPAVDQGG VNGHRYPGAG VVIKQEQTDF AYDSDVTGCA SMYLHTEGFS GPSPGDGAMG YGYEKPLRPF PDDVCVVPEK FEGDIKQEGV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
ETV4: ets variant gene 4 (E1A enhancer binding protein, E1AF), also known as PEA3. Several members of the Ets gene family are known to encode sequencespecific DNA binding proteins. These include mouse PU.1, mouse and human Ets-1, Drosophila E74, chicken and human Ets-2 and rat GABP-a. Each of these proteins recognizes similar motifs in DNA that share a centrally located 5′-GGAA-3′ element. For instance, PEA3 binds the motif 5′-AGGAAG-3′ (the PEA-3 motif), but does not bind to the sequence 5′-AGGAAC-3′, recognized by PU.1, although PU.1 binds equally well to both sequences. It appears thatall of the Ets proteins recognize the same central core sequence but that each protein interacts with unique sequences that flank this core. PEA3 is expressed at readily detectable levels in cells of epithelial and fibroblastic origin but is not expressed in hematopoietic cells. This is in contrast to other members of the Ets gene family, such as Ets-1, Ets-2 and Fli-1, each of whichis expressed primarily in cells of hematopoietic origin.
Especificaciones
Especificaciones
| Antígeno | ETV4 |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.05 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | ETV4 |
| N.º de referencia del gen | P43268 |
| Alias de gen | ETS translocation variant 4; Adenovirus E1A enhancer-binding protein; E1A-F; E1AF; ETV4 |
| Símbolos de los genes | ETV4 |
| Mostrar más |
Research Use Only
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?