missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ETV4 Monoclonal Antibody (1A2G3)
GREENER_CHOICE

Código de producto. 11385393
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
11385393 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11385393

Marca: Invitrogen™ MA515424

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA5-15424 targets ETV4 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15424 immunogen is purified recombinant fragment of human ETV4 (aa50-109) expressed in E. Coli. MA5-15424 detects ETV4 which has a predicted molecular weight of approximately 54kDa.

ETV4: ets variant gene 4 (E1A enhancer binding protein, E1AF), also known as PEA3. Several members of the Ets gene family are known to encode sequencespecific DNA binding proteins. These include mouse PU. 1, mouse and human Ets-1, Drosophila E74, chicken and human Ets-2 and rat GABP-a. Each of these proteins recognizes similar motifs in DNA that share a centrally located 5'-GGAA-3' element. For instance, PEA3 binds the motif 5'-AGGAAG-3' (the PEA-3 motif), but does not bind to the sequence 5'-AGGAAC-3', recognized by PU. 1, although PU. 1 binds equally well to both sequences. It appears that all of the Ets proteins recognize the same central core sequence but that each protein interacts with unique sequences that flank this core. PEA3 is expressed at readily detectable levels in cells of epithelial and fibroblastic origin but is not expressed in hematopoietic cells. This is in contrast to other members of the Ets gene family, such as Ets-1, Ets-2 and Fli-1, each of which is expressed primarily in cells of hematopoietic origin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno ETV4
Aplicaciones ELISA, Western Blot
Clasificación Monoclonal
Clon 1A2G3
Concentración Conc. Not Determined
Conjugado Unconjugated
Formulación Ascites with 0.03% sodium azide
génica ETV4
N.º de referencia del gen P43268
Alias de gen Adenovirus E1A enhancer-binding protein; AW414408; E1A F; E1AF; E1A-F; ETS translocation variant 4; ets variant 4; ets variant gene 4 (E1A enhancer binding protein, E1AF); ets variant gene 4 (E1A enhancer-binding protein, E1AF); ETS variant protein 4; Etv4; EWS protein/E1A enhancer binding protein chimera; PEA3; Pea-3; PEA3 protein; PEAS3; Polyomavirus enhancer activator 3; POLYOMAVIRUS ENHANCER ACTIVATOR 3 (PEA3 PROTEIN) (ETS TRANSLOCATION VARIANT 4); polyomavirus enhancer activator 3 homolog; polyomavirus enhancer activator-3; Protein PEA3
Símbolos de los genes ETV4
Especie del huésped Mouse
Inmunógeno Purified recombinant fragment of human ETV4 (aa50-109) expressed in E. Coli.
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2118
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.