Learn More
TERUMO SR STRAIGHT FEP Intravenous Catheters SURFLO™
Descripción
- The Surflo I.V. Catheters are made out of medical grade EFTE
- The materials of the Surflo™ I.V. Catheters are carefully selected to ensure a stable form during punctures under difficult conditions
- Short PIVC with an ETFE catheter tube and stainless steel internal needle
- Precisely tapered and siliconised catheter for smooth advancement of the catheter
- Catheter with X-ray contrast strips for higher patient safety when placement goes wrong
- The flashback chamber confirms needle tip entry into the vessel
- The large range in catheters and related obturators allows most appropriate choice of product for a wide range of applications
Especificaciones
Especificaciones
| Material | ETFE, Stainless Steel (Needle) |
| Color | Orange |
| Calibre de aguja | 14 G |
| Sin látex | Yes |
| Longitud de la aguja (imperial) | 2.5 in. |
| Longitud (métrico) | 64 mm |
| Tipo de producto | Catéter Surflo |
| Desechables | Single-use |
| Tipo de conexión | Leur Slip |
| Esterilidad | Sterile |