missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ESI 05

Click to view available options
Cantidad:
5 mg
25 mg
Tamaño de la unidad:
25 miligramos
5 miligramos
Description
ESI 05 is an EPAC2 inhibitor. Inhibits cAMP binding to EPAC2 and cAMP mediated EPAC2 guanine nucleotide exchange factor (GEF) activity (IC50 = 0.4 μM).
Specifications
Specifications
| Nombre del producto químico o material | 1,3,5-Trimethyl-2-[(4-methylphenyl)sulfonyl]benzene |
| CAS | 5184-64-5 |
| Cantidad | 25 mg |
| Fórmula molecular | C16H18O2S |
| Pureza | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction