Learn More
STEMCELL Technologies Erythropoietin (EPO) ELISA Kit
Código de producto. 17179991
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1 Kit
Tamaño de la unidad:
1 set
17179991 1 Kit 1 set
1 options
Código de producto. 17179991
Marca: STEMCELL Technologies 01630
Detection and measurement of human erythropoietin
  • The glycoprotein EPO is the main physiological regulator of red blood cell formation
  • The EPO ELISA Kit is a rapid 3-step enzyme-linked immunosorbent assay (ELISA) designed for the quantitative measurement of natural and recombinant human EPO
  • The assay uses two monoclonal antibodies raised against human urinary EPO
  • These antibodies bind two non-overlapping epitopes on the EPO polypeptide and show high-affinity binding to both natural and recombinant EPO
  • The range of detection for the kit is 1.6 - 100 mU/mL
  • The EPO ELISA Kit has an assay time of approximately 3 hours
Especificaciones
Descripción Other
Para utilizar con (aplicación) ELISA
Cantidad 1 Kit