missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
This antibody can recognize both ERK1 and ERK2 with the molecular mass of 38-43 kDa.
MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. The p44/42 MAPK (Erk1/2) are members of mitogen-activated kinases (MAPKs) of serine/threonine protein kinases. Phospho ERK1/2 can be activated by a range of extracellular stimuli, such as mitogen, growth factors, neurotransmitters, chemokines, and cytokines, through receptor tyrosine kinases (RTK), G protein-coupled receptors (GPCRs), or protein kinase C (PKC). Upstream MEK1 and MEK2 activate p44 and p42 (phospho ERK1/2) through phosphorylation of activation loop residues Thr202/Tyr204 and Thr185/Tyr187, respectively. p44/42 (ERK1/2) are negatively regulated by a family of dual-specificity (Thr/Tyr) MAPK phosphatases, known as DUSPs or MKPs, along with MEK inhibitors, such as U0126 and PD98059.
Especificaciones
Especificaciones
| Antígeno | ERK1/2 |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.22 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | MAPK1 |
| N.º de referencia del gen | P27361, P28482, P63085, Q63844 |
| Alias de gen | ERK, ERK 1, ERK1, ERK1/2, ERT2, HS44KDAP, HUMKER1A, Insulin stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MAPK3, p44 ERK1, p44 MAPK, P44ERK1, P44MAPK, PRKM3 |
| Símbolos de los genes | MAPK1, MAPK3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?