missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ErbB3 Monoclonal Antibody (RTJ1)
GREENER_CHOICE

Código de producto. 11511420
Change view
Click to view available options
Cantidad:
250 μL
Tamaño de la unidad:
250 microlitros
Código de producto. Cantidad unitSize
11511420 250 μL 250 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11511420

Marca: Invitrogen™ MA122819

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Formalin-fixed paraffin-embedded tissues require antigen retrieval prior to immunostaining.

ErbB3 (c-erbB-3/HER-3) is a member of the type I family of growth factor receptors and binds to ligands in the heregulin family. ErbB3 is over-expressed in a variety of tumors in the prostate, bladder, breast, stomach, pancreas, and colon. Heregulin and EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents. The ErbB3 gene is located on the long arm of human chromosome 12 (12q13) and is transcribed into a 6. 2kb mRNA which is translated into a 160/180 kDa glycoprotein. The ErbB3 gene encodes a transmembrane receptor tyrosine kinase due to alternative splicing and forms heterodimers with other EGF receptor family members that have kinase activity. Heterodimerization leads to the activation of pathways that lead to cell proliferation or differentiation. Alternate transcriptional splice variants encoding different isoforms of ErbB3 have been characterized. One ErbB3 isoform lacks the intermembrane region, is secreted outside the cell, and acts to modulate the activity of the membrane-bound form. Additional splice variants have also been reported, but they have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno ErbB3
Aplicaciones Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon RTJ1
Concentración 0.1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
génica ERBB3
N.º de referencia del gen P21860
Alias de gen avian erythroblastosis oncogene B 3; avian erythroblastosis oncogene B 3 receptor; C76256; c-erbB3; c-erbB-3; C-errB-3; erb-b2 receptor tyrosine kinase 3; ERBB3; Erbb-3; Erbb3r; erbB3-S; glial growth factor receptor; HER3; human epidermal growth factor receptor 3; LCCS2; MDA BF 1; MDA-BF-1; MGC88033; nuc-ErbB3; p180 ErbB3; p180-ErbB3; p45 sErbB3; p45-sErbB3; p85 sErbB3; p85-sErbB3; proto-oncogene-like protein c-ErbB-3; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-3; receptor tyrosine-protein kinase erbB-3; LOW QUALITY PROTEIN: receptor tyrosine-protein kinase erbB-3; tyrosine kinase-type cell surface receptor HER3; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Símbolos de los genes ERBB3
Especie del huésped Mouse
Inmunógeno Synthetic peptide corresponding to a site on the cytoplasmic domain of the human c-erbB-3 oncoprotein.
Método de purificación Purified
Cantidad 250 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2065
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgM
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.