missing translation for 'onlineSavingsMsg'
Learn More

ERBB2 Mouse anti-Human, Clone: ERB2/776, Abnova™

Código de producto. 16061796
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
16061796 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16061796

Marca: Abnova MAB13268.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse monoclonal antibody raised against partial recombinant human ERBB2.

This gene encodes a member of the epidermal growth factor (EGF) receptor family of receptor tyrosine kinases. This protein has no ligand binding domain of its own and therefore cannot bind growth factors. However, it does bind tightly to other ligand-bound EGF receptor family members to form a heterodimer, stabilizing ligand binding and enhancing kinase-mediated activation of downstream signalling pathways, such as those involving mitogen-activated protein kinase and phosphatidylinositol-3 kinase. Allelic variations at amino acid positions 654 and 655 of isoform a (positions 624 and 625 of isoform b) have been reported, with the most common allele, Ile654/Ile655, shown here. Amplification and/or overexpression of this gene has been reported in numerous cancers, including breast and ovarian tumors. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized. [provided by RefSeq]

Especificaciones

Antígeno ERBB2
Aplicaciones Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Clasificación Monoclonal
Clon ERB2/776
Conjugado Unconjugated
Descripción Mouse monoclonal antibody raised against partial recombinant human ERBB2.
Dilución Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulación In 10mM PBS.
Alias de gen CD340/HER-2/HER-2/neu/HER2/NEU/NGL/TKR1
Símbolos de los genes ERBB2
Especie del huésped Mouse
Inmunógeno Recombinant protein corresponding to intracellular domain of human ERBB2.
Método de purificación Protein A/G purification
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2064
Especificidad de la prueba This monoclonal antibody is specific to ERBB2 and shows minimal cross-reaction with other members of the family.
Especies diana Human
Contenido y almacenamiento Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.