missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPS8L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA560413
This item is not returnable.
View return policy
Description
Immunogen sequence: PKPSAKSIYE QRKRYSTVVM ADVSQYPVNH LVTFCLGEDD GVHTVEDASR KLAVMDSQGR VWAQEMLLRV SPDHVTLLDP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.
Eps8L1 (EPS8-like 1), also known as DRC3 or EPS8R1, is a 723 amino acid protein that localizes to the cytoplasm and belongs to the Eps8 (epidermal growth factor receptor pathway substrate 8) family. Expressed in placental tissue, Eps8L1 functions to stimulate the guanine exchange activity of Sos 1 (son of sevenless homolog 1), a protein that promotes the exchange of Ras-bound GDP for GTP. Additionally, Eps8L1 is thought to associate with Actin and, via this association, may play a role in membrane ruffling and remodeling of the Actin cytoskeleton. Through its ability to regulate protein activation and cytoskeleton dynamics, Eps8L1 may participate in cell growth and differentiation events within the cell. Eps8L1 contains one SH3 domain and is expressed as four isoforms due to alternative splicing events.
Specifications
| EPS8L1 | |
| Polyclonal | |
| Unconjugated | |
| EPS8L1 | |
| 2310051G19Rik; 4632407K17Rik; AW060268; DRC3; Epidermal growth factor receptor kinase substrate 8-like protein 1; epidermal growth factor receptor pathway substrate 8-related protein 1; EPS8 like 1; Eps8l1; EPS8-like 1; EPS8-like protein 1; Eps8r1; EPS8-related protein 1; PP10566 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54869 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TE68 | |
| EPS8L1 | |
| Recombinant protein corresponding to Human EPS8L1. Recombinant protein control fragment (Product #RP-99081). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction