Learn More
Invitrogen™ EPS8 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Marca:  Invitrogen™ PA518196
Código de producto. 11376163
  • 532.00€ / 100 microgramos
Descripción
This antibody is predicted to react with canine, human, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene encodes a member of the EPS8 family. This protein contains one PH domain and one SH3 domain. It functions as part of the EGFR pathway, though its exact role has not been determined. Highly similar proteins in other organisms are involved in the transduction of signals from Ras to Rac and growth factor-mediated actin remodeling. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
EPS8
Polyclonal
Unconjugated
EPS8
AI042819; AW261790; AW545405; DFNB102; epidermal growth factor receptor kinase substrate 8; Epidermal growth factor receptor kinase substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8; epidermal growth factor receptor pathway substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8-related protein 2; EPS8; EPS8 related protein 2; Eps8l2; EPS8-like 2; EPS8-like protein 2; Eps8r2; EPS8-related protein 2
Goat
Ammonium sulfate precipitation
RUO
13860, 2059
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q08509, Q12929
EPS8
Synthetic peptide sequence (SGVESFDEGSSH) corresponding to the C-terminus amino acids of EPS8 (aa 811-822).
100 μg
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones