missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epo Monoclonal Antibody (Epo2)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA123056
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The predicted molecular weight for this product is 21 kDa.
Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.
Especificaciones
| Epo | |
| Monoclonal | |
| 2.7 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P01588 | |
| EPO | |
| Recombinant full length protein (Human). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| Epo2 | |
| Unconjugated | |
| EPO | |
| EP; EPO; Epoetin; erythropoietin; MGC138142; MVCD2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 2056 | |
| 4°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido