Learn More
Ephrin B2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26533-1-AP-20UL
Descripción
The Ephrin B2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Ephrin B2. This antibody recognizes human, mouse antigen. The Ephrin B2 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
| Ephrin B2 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| EFNB2, ephrin B2, EPLG5, Htk L, HTK ligand, HTKL, LERK 5, LERK5 | |
| 20 μL | |
| 1948 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |