missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EphB4 Monoclonal Antibody (7H4A6)

Descripción
MA5-15415 targets EphB4 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15415 immunogen is purified recombinant fragment of EphB4 (aa562-612)expressed in E. Coli.. MA5-15415 detects EphB4 which has a predicted molecular weight of approximately 108kDa.
EPHB4 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by EphB4 binds to ephrin-B2 and plays an essential role in vascular development.
Especificaciones
Especificaciones
| Antígeno | EphB4 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 7H4A6 |
| Concentración | Conc. Not Determined |
| Conjugado | Unconjugated |
| Formulación | Ascites with 0.03% sodium azide |
| génica | EPHB4 |
| N.º de referencia del gen | P54760 |
| Alias de gen | AI042935; Developmental kinase 2; EPH receptor B4; Ephb4; ephrin receptor EphB4; ephrin type-B receptor 4; Hepatoma transmembrane kinase; HTK; Mdk2; mDK-2; MYK1; soluble EPHB4 variant 1; soluble EPHB4 variant 2; soluble EPHB4 variant 3; TYRO11; Tyrosine kinase MYK-1; tyrosine-protein kinase receptor HTK; tyrosine-protein kinase TYRO11 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?