missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPCR (CD201) Rat, anti-Mouse, PE, Clone: RMEPCR1560 (1560), STEMCELL Technologies
Descripción
- The RMEPCR1560 (1560) antibody reacts with the endothelial protein C receptor (EPCR or CD201), an ∼25 kDa type I transmembrane glycoprotein expressed by endothelial cells, subsets of hematopoietic stem cells (HSCs) and dendritic cells, and several malignant cell lines
- It is also found in a soluble form in plasma
- EPCR exhibits homology with the MHC class 1/CD1 protein family
- EPCR binds protein C and activated protein C, thus augmenting protein C activation by the thrombin-thrombomodulin complex and regulating blood coagulation and inflammation
- EPCR protein expression has been detected on ∼1.5% of mouse bone marrow cells
- Purified EPCR+ cells are highly enriched for HSC activity, as evidenced by high in vivo repopulation activity
- Moreover, EPCR expression is associated with the stem cell activity of bone marrow cell populations isolated using conventional markers, indicating the usefulness of EPCR as a single marker for the identification of mouse HSCs
Especificaciones
Especificaciones
| Antígeno | EPCR (CD201) |
| Aplicaciones | Immunohistochemistry, Western Blot, Flow Cytometry, Functional Assay |
| Clasificación | Monoclonal |
| Clon | RMEPCR1560 (1560) |
| Conjugado | PE |
| Especie del huésped | Rat |
| Cantidad | 100 μg |
| Primario o secundario | Primary |
| Especies diana | Mouse |
| Isotype | IgG2b κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?