Learn More
Invitrogen™ Eotaxin 2 Polyclonal Antibody

Goat Polyclonal Antibody
Marca: Invitrogen™ PA125517
Descripción
For neutralization, use at a concentration of 7-8 μg/mL to achieve half of the neutralizing dose (ND50) of a 100.0 ng/mL concentration.
Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.
Especificaciones
| Eotaxin 2 | |
| Polyclonal | |
| Unconjugated | |
| CCL24 | |
| CC chemokine CCL24; C-C motif chemokine 24; C-C motif chemokine ligand 24; Ccl24; chemokine (C-C motif) ligand 24; Ckb-6; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin2; eotaxin-2; eotaxin-2-like protein; MPIF2; MPIF-2; Myeloid progenitor inhibitory factor 2; Scya24; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; small-inducible cytokine A24 | |
| Goat | |
| Affinity chromatography | |
| RUO | |
| 56221 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Functional Assay, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9JKC0 | |
| CCL24 | |
| Recombinant mouse Eotaxin 2. | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.