missing translation for 'onlineSavingsMsg'
Learn More

PeproTech Eotaxin 2 Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Código de producto. 17864633
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
Código de producto. Cantidad unitSize
17864633 1 mg 1 miligramo
17844063 50 μg 50 microgramos
17833863 100 μg 100 microgramos
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17864633

Marca: PeproTech 500P103G1MG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody

AA Sequence of recombinant protein: VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVA. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human Eotaxin-2 (CCL24). Anti-Human Eotaxin-2 (CCL24)-specific antibody was purified by affinity chromatography employing an immobilized Human Eotaxin-2 (CCL24) matrix. Sandwich ELISA: To detect Human Eotaxin-2 (CCL24) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Eotaxin-2 (CCL24) (500-P103GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Eotaxin-2 (CCL24). Western Blot: To detect Human Eotaxin-2 (CCL24) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Eotaxin-2 (CCL24) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Eotaxin 2
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica CCL24
N.º de referencia del gen O00175
Alias de gen CC chemokine CCL24; C-C motif chemokine 24; C-C motif chemokine ligand 24; Ccl24; chemokine (C-C motif) ligand 24; Ckb-6; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin2; eotaxin-2; eotaxin-2-like protein; MPIF2; MPIF-2; Myeloid progenitor inhibitory factor 2; Scya24; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; small-inducible cytokine A24
Símbolos de los genes CCL24
Especie del huésped Goat
Inmunógeno E.coli-derived Recombinant Human Eotaxin-2 (CCL24).
Método de purificación Antigen affinity chromatography
Cantidad 1 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6369
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.