missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Enterobacteriaciae ECA Monoclonal Antibody (1055)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA533256
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
For ELISA sandwich assay, employ MA5-33256 as biotin conjugate.
Enterobacterial common antigen (ECA) is a family-specific surface antigen shared by all members of the Enterobacteriaceae and is restricted to this family. ECA is found in freshly isolated wild-type strains as well as in laboratory strains like Escherichia coli K-12. The family specificity of ECA can be used for taxonomic and diagnostic purposes. ECA is located in the outer leaflet of the outer membrane.
Especificaciones
| Enterobacteriaciae ECA | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| 1 mL | |
| Primary | |
| 4°C, do not freeze | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 1055 | |
| Unconjugated | |
| Enterobacteria | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido