Learn More
Lonza™ Endothelial Basal Medium
Código de producto. 11675420
Click to view available options
:
Product Type: Endothelial Basal Medium
Código de producto. 11675420
Marca: Lonza™ CC3121
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Endothelial Basal Medium is part of a collection of tools optimized to grow certain cell types.
Media products are supplied in several formats:
  • Basal Medium -– contains no growth factors, cytokines, or supplements
  • SingleQuots™ Kit – growth factors, cytokines, and supplements
  • BulletKit™ – basal medium and SingleQuots™ Kit packaged together
The following Endothelial Cell Growth Medium BulletKits™ are available:
  • EGM™-2 BulletKit™ – Optimized for certain HUVECs, Aortic and Pulmonary artery endothelial cells. Contains 2% FBS and VEGF for rapid proliferation. Does not contain BBE.
  • EGM™ BulletKit™ – Optimized for HUVECs cat. no. CC-2517 and CC-2519, contains 2% FBS and BBE. Does not contain exogenous VEGF.
  • New! EGM™-Plus BulletKit™ – Enhanced version of EGM™ Medium. Optimized for HUVECs cat. no. CC-2935. Contains 2% FBS and BBE. Does not contain exogenous VEGF. Faster proliferation than HUVECs isolated in EGM™ Medium.
  • EGM™-2MV BulletKit™ – Coronary and Iliac artery, human microvascular endothelial cells, contains 5% FBS and VEGF for rapid proliferation. Does not contain BBE.
  • EGM™ -MV BulletKit™ – Bovine Microvascular Cells, contains 5% FBS and BBE. Does not contain exogenous VEGF.
Información de órdenes
dimTipProducto
TRUSTED_SUSTAINABILITY
Especificaciones
Tipo de producto Basal Medium