missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enasidenib mesylate, MedChemExpress
Descripción
Enasidenib mesylate is a first-in-class, oral, potent, reversible, selective inhibitor of the IDH2 mutant enzymes.
Especificaciones
Specifications
| Nombre del producto químico o material | Enasidenib mesylate |
| CAS | 1650550-25-6 |
| Color | Blanco |
| Notas de grado de pureza | Research |
| Fórmula molecular | C20H21F6N7O4S |
| Cantidad | 50 mg |
| Sinónimo | AG-221 mesylate |
| Información de solubilidad | DMSO : ≥ 100 mg/mL (175.60 mM) |
| SMILES | CS(=O)(O)=O.CC(O)(C)CNC1=NC(C2=NC(C(F)(F)F)=CC=C2)=NC(NC3=CC(C(F)(F)F)=NC=C3)=N1 |
| Peso molecular (g/mol) | 569.48 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?