missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EMX2 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15921405
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
0.10 miligramo
Código de producto. Cantidad unitSize
15921405 100 μg 0.10 miligramo
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15921405

Marca: Invitrogen™ PA572743

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

EMX2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a homeobox-containing transcription factor that is the homolog to the 'empty spiracles' gene in Drosophila. Research on this gene in humans has focused on its expression in three tissues: dorsal telencephalon, olfactory neuroepithelium, and urogenetial system. It is expressed in the dorsal telencephalon during development in a low rostral-lateral to high caudal-medial gradient and is proposed to pattern the neocortex into defined functional areas. It is also expressed in embryonic and adult olfactory neuroepithelia where it complexes with eukaryotic translation initiation factor 4E (eIF4E) and possibly regulates mRNA transport or translation. In the developing urogenital system, it is expressed in epithelial tissues and is negatively regulated by HOXA10. Alternative splicing results in multiple transcript variants encoding distinct proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno EMX2
Aplicaciones ELISA, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.02% sodium azide
génica EMX2
N.º de referencia del gen Q04743
Alias de gen empty spiracles homeobox 2; empty spiracles homolog 2; Empty spiracles-like protein 2; Emx2; Emx-2; Homeobox protein EMX2; Pdo; RGD1564797
Símbolos de los genes EMX2
Especie del huésped Rabbit
Inmunógeno EMX2 antibody for an 18 amino acid synthetic peptide near the center of human EMX2. The immunogen is within amino acids 60 - 110 of EMX2.
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2018
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.