Learn More
Kartell Embedding Rings
Descripción
- Embedding supports suitable for the processing and the histological inclusion of paraffin pieces
- The inclusion rings can use the same microtome support (instrument used to make the histological sections of samples) of the inclusion cassettes
- They can be set up on every kind of microtome thanks to their structure and to the possibility to obtain uniform paraffin blocks.
- Embedding rings is an easy-filling and, after cutting, is always ready for following sections
Especificaciones
Especificaciones
| Tipo de producto | Embedding Ring |
| Material | Plástico |
| Color | Verde |
| Longitud (métrico) | 41 mm |
| Anchura (métrico) | 28 mm |
| Longitud (imperial) | 1.61 in. |
| Anchura (imperial) | 1.1 in. |
| Fondo (métrico) | 11 mm |
| Fondo (imperial) | 0.43 in. |
| Desechables | Single-use |