missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-83819
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ELP2 Polyclonal specifically detects ELP2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
| ELP2 | |
| Polyclonal | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| elongation protein 2 homolog (S. cerevisiae), elongator complex protein 2, elongator protein 2, FLJ10879, SHINC-2, signal transducer and activator of transcription 3 interacting protein 1, signal transducer and activator of transcription interacting protein 1, STAT3-interacting protein 1, STATIP1, StIP, stIP1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55250 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ELP2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ECDSTDDCIEHNIGPCSSVLDVGGAVTAVSVCPVLHPSQRYVVAVGLECGKICLYTWKKTDQVPEINDWTHCVETSQSQSHTLAIRKLCWKNCSGKTEQKEAEGAEWLHFASCGEDHTVKIHRVNKCA | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido