missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELOVL6 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 10813461
Change view
Click to view available options
Cantidad:
400 μL
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
10813461 400 μL 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10813461 Proveedor Invitrogen™ N.º de proveedor PA513455

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

This antibody is predicted to react with chicken and rat based on sequence homology.

Lipogenesis is a key event in the energy storage system and is controlled by the transcription factor sterol regulatory element-binding protein (SREBP)-1. Elongation of very long chain fatty acids protein 6 (ELOVL6) is a member of fatty acyl-CoA elongase gene family that converts palmitic to stearic acid and it has been shown to be a target of SREBP-1, playing an important role in de novo synthesis of long-chain saturated and monosaturated fatty acids in conjunction with fatty acid synthase and stearoyl-CoA desaturase ELOVL6 was predicted to be important for tissue fatty acid composition. Recent studies suggest that inhibition of this elongase could be a new therapeutic approach for ameliorating insulin resistance, diabetes and cardiovascular risks, even in the presence of a continuing state of obesity.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno ELOVL6
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 2 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.09% sodium azide; pH 7.4
génica ELOVL6
N.º de referencia del gen Q920L5, Q9H5J4
Alias de gen 3-keto acyl-CoA synthase Elovl6; C77826; Elongation of very long chain fatty acids protein 6; ELOVL FA elongase 6; ELOVL family member 6, elongation of long chain fatty acids; ELOVL family member 6, elongation of long chain fatty acids (FEN1/Elo2, SUR4/Elo3-like, yeast); ELOVL family member 6, elongation of long chain fatty acids (yeast); ELOVL fatty acid elongase 6; ELOVL6; FACE; FAE; fatty acid elongase 2; Fatty acyl-CoA elongase; hELO2; Lce; Lce2; long chain fatty acid elongase; long chain fatty acyl elongase; long-chain fatty-acyl elongase; Masr; Myelin-associated SUR4 protein; rELO2; Very long chain 3-ketoacyl-CoA synthase 6; very long chain 3-oxoacyl-CoA synthase 6; very-long-chain 3-oxoacyl-CoA synthase 6
Símbolos de los genes ELOVL6
Especie del huésped Rabbit
Inmunógeno KLH conjugated synthetic peptide between 32-58 amino acids from the N-terminal region of human ELOVL6.
Método de purificación Ammonium sulfate precipitation, Size-exclusion - Dialysis
Cantidad 400 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 170439, 79071
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.