Learn More
EIF4A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16808253 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16808253 20 μL 20 microlitros
16898243 150 μL 150 microlitros
2 options
Código de producto. 16808253
Marca: Proteintech 112801AP20UL
Rabbit Polyclonal Antibody
eIF4A2 is a 407 amino acid ATP-dependent RNA helicase belonging to the DEAD box helicase family. It contains one helicase ATP-binding domain and a helicase C-terminal domain. eIF4A2, a subunit of the eIF4F complex, acts as a protein-synthesis initiation factor and thus plays an important role in the binding of mRNA to the 43S pre-initiation complex of ribosomes. Human eIF4A2 interacts with the NS5B protein of Hepatitis C virus (HCV) and facilitates the genomic RNA synthesis of NS5B protein by unwinding the secondary structure of the HCV genome. It is ubiquitously expressed in most of normal tissues, but in variable amounts.
Especificaciones
Antígeno EIF4A2
Aplicaciones Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.13 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica EIF4A2
N.º de referencia del gen P10630, Q14240, Q5RKI1
Alias de gen BM 010, DDX2B, eIF 4A II, EIF4A, eIF4A II, EIF4A2, EIF4F
Símbolos de los genes EIF4A2
Especie del huésped Rabbit
Inmunógeno EIF4A2 Fusion Protein Ag1810
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 13682, 1974, 303831
Especies diana Human, Rat, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG