missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3M Mouse anti-Human, Clone: 2A12B2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66025-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a protein that is part of the eurkaryotic translation initiation factor 3 complete (eIF-3) required for protein synthesis. Elevated levels of the encoded protein are present in cancer cell lines. Inactivation of the encoded protein has been shown to interfere with translation of herpes virus mRNAs by preventing the association of mRNAs with the ribosomes. A pseudogene of this gene is located on the X chromosome.Especificaciones
| EIF3M | |
| Monoclonal | |
| 0.68 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7L2H7 | |
| EIF3M | |
| EIF3M Fusion Protein Ag17965 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 2A12B2 | |
| Unconjugated | |
| EIF3M | |
| B5, EIF3M, Fetal lung protein B5, GA17, hfl B5, HFLB5, PCID1, PNAS 125 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10480 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido