missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF3h Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15732631 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
15732631 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15732631

Marca: Invitrogen™ PA554699

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: QQQQKHQYQQ RRQQENMQRQ SRGEPPLPEE DLSKLFKPPQ PPARMDSLLI AGQINTYCQN IKEFTAQNLG KLFMAQALQE YNN Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 96%.

eIF3H is a component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno eIF3h
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica Eif3h
N.º de referencia del gen O15372, Q6P9U8, Q91WK2
Alias de gen 1110008A16Rik; 40 kDa; 40kD; 9430017H16Rik; eIF-3 gamma; eIF3 p40 subunit; eIF3 p40 subunit {ECO:0000255; eIF-3-gamma; eIF3-gamma; EIF3H; eIF3h {ECO:0000255; eIF3p40; eIF3-p40; EIF3S3; eukaryotic translation initiation factor 3 subunit 3; eukaryotic translation initiation factor 3 subunit 3 {ECO:0000255; eukaryotic translation initiation factor 3 subunit 3 gamma; eukaryotic translation initiation factor 3 subunit H; eukaryotic translation initiation factor 3 subunit H {ECO:0000255; eukaryotic translation initiation factor 3 subunit H-like protein; eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD); eukaryotic translation initiation factor 3, subunit 3 (gamma); eukaryotic translation initiation factor 3, subunit 3 (gamma, 40kD); eukaryotic translation initiation factor 3, subunit 3 gamma, 40kDa; eukaryotic translation initiation factor 3, subunit H; eukaryotic translation initiation factor subunit H-like protein; HAMAP-Rule:MF_03007}; MGC102958
Símbolos de los genes Eif3h
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human eIF3h. Recombinant protein control fragment (Product #RP-93817).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 299899, 68135, 8667
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.