Learn More
EIF3D Rabbit anti-Human, Mouse, Yeast, Polyclonal, Proteintech
Código de producto. 16856463 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16856463 20 μL 20 microlitros
16846463 150 μL 150 microlitros
2 options
Código de producto. 16856463
Marca: Proteintech 102191AP20UL
Rabbit Polyclonal Antibody
eIF3D is a protein belonging to the largest of the eukaryotic translation initiation factor family, eukaryotic translation initiation factor-3 (eIF3). eIF3 is a multiprotein complex composed of at least 13 different subunits. eIF3D binds to the 40S ribosomal subunit and thus helps in maintaining the 40S and 60S ribosomal subunits in a dissociated state. An important role of eIF3D is the formation of the 40S initiation complex by interacting with the ternary complex of eIF2/GTP/methionyl-tRNA, and promotes mRNA binding. eIF3D is the major RNA binding subunit of the eIF3 complex. It associates with the subunit p170 of eIF-3. Ubiquitous expression of eIF3D is seen in most of the tissues and it is phosphorylated upon DNA damage, likely by ATM or ATR.
Especificaciones
Antígeno EIF3D
Aplicaciones Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.29 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica EIF3D
N.º de referencia del gen O15371, O70194
Alias de gen eIF 3 zeta, eIF3 p66, eIF3 zeta, EIF3D, EIF3S7
Símbolos de los genes Eif3d
Especie del huésped Rabbit
Inmunógeno EIF3D Fusion Protein Ag0268
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 55944, 8664
Especies diana Mouse, Yeast, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG