Learn More
EGFR (Epidermal Growth Factor Receptor) Mouse, Clone: GFR/2341, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 1956-MSM11-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
EGFR (Epidermal Growth Factor Receptor) | |
Cell Surface | |
GFR/2341 | |
Unconjugated | |
Mouse | |
∽170kDa (wild type) and ∽145kDa (vIII variant) | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
1956 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human EGFR protein (apprx. aa 344-492) | |
A431 cells. Placenta, Breast, Colon or Bladder cancer | |
100 μg | |
Primary | |
IgG2b κ |