missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: PCSPMPRSPE ECRRLRTCSE CLARHPRTLQ PGDGEASTPR CKWCTNCPEG ACIGRNGSCT SENDCRINQR EVFWAGNCSE AACGAADCEQ CTREGKCMWT RQFKRTGETR RILSVQPTYD WTCFSHSLLN VS Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
EGFL4 (epidermal growth factor-like protein 4), also known as MEGF8 (multiple epidermal growth factor-like domains protein 8) or C19orf49, is a 2,845 amino acid single-pass type I membrane protein. EGFL4 contains 2 CUB domains, 5 EGF-like domains, 12 Kelch repeats, 4 laminin EGF-like domains and 7 PSI domains. Existing as two alternatively spliced isoforms, the gene encoding EGFL4 maps to human chromosome 19q13.2. Chromosome 19 consists of approximately 63 million bases, makes up over 2% of human genomic DNA, and is recognized for having the greatest gene density of the human chromosomes. It is the genetic home for a number of immunoglobulin superfamily members including the killer cell and leukocyte Ig-like receptors.
Especificaciones
Especificaciones
| Antígeno | EGFL4 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.05 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | MEGF8 |
| N.º de referencia del gen | Q7Z7M0 |
| Alias de gen | AW049492; b2b1702Clo; b2b288Clo; C19orf49; CRPT2; EGFL4; EGF-like domain-containing protein 4; EGF-like protein 4; EGF-like-domain, multiple 4; Epidermal growth factor-like protein 4; HBV pre-s2 binding protein 1; HBV pre-S2-binding protein 1; hepatitis B virus pre-S2-binding protein 1; KIAA0817; m687Ddg; Megf8; mKIAA0817; multiple EGF like domains 8; multiple EGF-like domain protein 4; multiple EGF-like domains protein 8; multiple EGF-like-domains 8; multiple epidermal growth factor-like domains protein 8; SBP1 |
| Símbolos de los genes | MEGF8 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
