missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGF Receptor Mouse anti-Human, BUV563, Clone: EMab-134, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 749748
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
EGF Receptor Monoclonal antibody specifically detects EGF Receptor in Human samples. It is validated for Flow Cytometry
Especificaciones
| EGF Receptor | |
| Monoclonal | |
| Brilliant Ultraviolet 563 | |
| EGFR; ERBB; ERBB1; HER1; mENA; PIG61 | |
| Human EGF Receptor extracellular domain Recombinant Protein | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| EMab-134 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido