Learn More
EEF1D Monoclonal Antibody (OTI6E1), TrueMAB™, OriGene
Descripción
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | EEF1D |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI6E1 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | EEF1D |
| N.º de referencia del gen | P29692 |
| Alias de gen | EF-1D, EF1D, FP1047 |
| Mostrar más |