Learn More
Invitrogen™ EED Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA584480
Descripción
Immunogen sequence: HPLLAVAGSR GIIRIINPIT MQCIKHYVGH GNAINELKFH PRDPNLLLSV SKDHALRLWN IQTDTLVAIF GGVEGHRDEV LSADYDLLGE KIMSCGMDHS LK Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
The EED protein (WAIT-1 or WD protein associated with integrin cytoplasmic tails-1), also called embryonic ectoderm development, is a member of the superfamily of WD-40 repeat proteins and widely conserved polycomb group (PcG) family of proteins. The polycomb group (PcG) is a large and evolutionarily conserved set of genes whose products act in multimeric complexes to modify histones, which are then thought to cause stable and heritable states of transcriptional repression. EED shuttles between the nucleus and the plasma membrane and can interact with the cytoplasmic tail of integrin Beta-7 subunit. EED exerted an antiviral activity at the late stage of HIV-1 replication.
Especificaciones
| EED | |
| Polyclonal | |
| Unconjugated | |
| EED | |
| EED; embryonic ectoderm development; Embryonic ectoderm development protein; ENSMUSG00000039373; HEED; l(7)5Rn; l7Rn5; lethal, Chr 7, Rinchik 5; lusk; Polycomb protein EED; WAIT1; WAIT-1; WD protein associating with integrin cytoplasmic tails 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75530 | |
| EED | |
| Recombinant protein corresponding to Human EED. Recombinant protein control fragment (Product #RP-103391). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.