missing translation for 'onlineSavingsMsg'
Learn More

EDA2R/TNFRSF27/XEDAR Antibody, CoraFluor™ 1, Novus Biologicals™

Código de producto. 30141017
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
30141017 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 30141017

Brand: Novus Biologicals (Bio-Techne) NBP176710CL1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EDA2R/TNFRSF27/XEDAR Polyclonal antibody specifically detects EDA2R/TNFRSF27/XEDAR in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno EDA2R/TNFRSF27/XEDAR
Aplicaciones ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado CoraFluor 1
Formulación PBS
Alias de gen ectodysplasin A2 receptor, EDA-A2 receptor, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor
Especie del huésped Rabbit
Inmunógeno Antibody was raised against recombinant human XEDAR.
Método de purificación Peptide affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 60401
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C in the dark. Do not freeze.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.