Learn More
Skytec™ Eco Bronze Gloves
GREENER_CHOICE
Código de producto. p-200110052
Change view
Click to view available options
Tamaño:
Pequeño
Mediano
Grande
XL
XXL
Tamaño de la unidad:
5 pares
Código de producto. Tamaño unitSize
30274611 Pequeño 5 pares
30274599 Mediano 5 pares
30274563 Grande 5 pares
30274537 XL 5 pares
30274638 XXL 5 pares
5 options
Código de producto. 30274611
Marca: Skytec™ SKG00052ED
Solo quedan null
Añadir a la cesta
Añadir a la cesta
A very flexible thermal glove with recycled PET liner and nitrile foam knuckle coating for work in oily and wet conditions. Touchscreen friendly and 360° water repellent treatment.
  • Insulated Glove with Recycled Polyester Liner
  • Soft and lightweight liner made of 50% recycled polyester and 50% virgin polyester
  • Flexible and durable nitrile foam knuckle coating guarantee a secure grip in wet and oily conditions
  • Light acrylic inner liner provides thermal insulation
  • 360° water repellent treatment to protect from liquids
  • Touchscreen capabilities to allow usage with screen devices
  • Close-fitting liner for a comfortable wearing experience
  • Elasticated wrist provides a secure fit
  • RCS (Recycled Claim Standard) certified
  • 1 pair = 1 500 mL (PET) plastic bottle
Especificaciones
Certificaciones/conformidad (Europa) CE II, UKCA, ANSI/ISEA 105-2016: Cut A2, EN 388:2016 + A1:2018 3331B, EN 21420:2020
Estructura Seamless Knit
Coating Material Nitrile Foam
Color Green, Black
Acabado exterior Rough
Hand Ambidextrous
Material Polyester
Tipo de producto Insulated Gloves
Tamaño Pequeño
Material de revestimiento Acrylic
Longitud (métrico) 240 mm