Learn More
Tocris Bioscience™ Ebselen
GREENER_CHOICE
Código de producto. p-5545776
Change view
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
18785666 10 mg 10 miligramos
18767426 50 mg 50 miligramos
2 options
Código de producto. 18785666
Marca: Tocris Bioscience™ 5245/10
Glutathione peroxidase mimic; peroxynitrite scavenger
Ebselen is a glutathione peroxidase mimic; peroxynitrite scavenger. Inhibits ferroptosis. Also inhibits lipoxygenase, cyclooxygenase, nitric oxide synthase, protein kinase C and H+/K+-ATPase activity. Inhibits the hepatic carcinogenic effects of aflatoxin B1. Antioxidant and anti-inflammatory. Selectively inhibits Gq protein signaling and enhances differentiation of brown adipocytes. Inhibits SARS-CoV-2 Mpro in vitro (IC50 = 0.67 μM in FRET assay)
TRUSTED_SUSTAINABILITY
Especificaciones
CAS 60940-34-3
Cantidad 10 mg
Fórmula molecular 2-Phenyl-1,2-benzisoselenazol-3(2 H)-one
Tipo de producto Ebselen
Pureza 98%
Peso molecular M.W. (Observed): 274.18
Inhibidores Ebselen
Para uso exclusivo en investigación