Learn More
STEMCELL Technologies EasySep™ Release Human CD4 Positive Selection Kit
Código de producto. 17177751
Encompass
Change view
Click to view available options
Cantidad:
1 x 109 Celdas
Tamaño de la unidad:
1 set
17177751 1 x 109 Celdas 1 set
1 options
Código de producto. 17177751
Marca: STEMCELL Technologies 17752
Immunomagnetic positive selection of particle-free human CD4+ cells
  • The EasySep™ Release Human CD4 Positive Selection Kit is designed to isolate CD4+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection
  • Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
  • Unwanted cells are simply poured off, while desired cells remain in the tube
  • Then, bound magnetic particles are removed from the EasySep™-isolated CD4+ cells, which are immediately available for downstream applications
  • Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands
  • The CD4 antigen is strongly expressed on helper T cells and is expressed more weakly on peripheral blood monocytes and tissue macrophages
Especificaciones
Tipo de célula T Cells, T Cells, CD4+
Cantidad 1 x 109 Celdas