Learn More
STEMCELL Technologies EasySep™ Red Blood Cell Lysis Buffer
Código de producto. 17108021
Encompass_Preferred
Change view
Click to view available options
Cantidad:
10 mL
100 mL
Tamaño de la unidad:
10 ml
100 ml
17108021 10 mL 10 ml
17138021 100 mL 100 ml
2 options
Código de producto. 17108021
Marca: STEMCELL Technologies 20110
Red blood cell lysis buffer, 10X concentrate
  • EasySep™ Red Blood Cell Lysis Buffer is for use as directed with EasySep™ or RoboSep™ Whole Blood kits for the lysis of red blood cells prior to separation from whole blood
Especificaciones
Descripción Chimerism; HLA; Immunology
Cantidad 10 mL