Learn More
STEMCELL Technologies EasySep™ Mouse APC Positive Selection Kit II
Código de producto. 17177731
Encompass
Change view
Click to view available options
Cantidad:
1 x 109 Celdas
Tamaño de la unidad:
1 set
17177731 1 x 109 Celdas 1 set
1 options
Código de producto. 17177731
Marca: STEMCELL Technologies 17667
Immunomagnetic positive selection of mouse cells labeled with APC-conjugated antibodies
  • The EasySep™ Mouse APC Positive Selection Kit II is designed to isolate cells that are labeled with APC (allophycocyanin)-conjugated antibodies
  • Desired cells are targeted with antibody complexes recognizing APC and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • This product replaces the EasySep™ Mouse APC Positive Selection Kit (Catalog #18452)
Especificaciones
Tipo de célula B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells
Para utilizar con (aplicación) Cell Isolation
Cantidad 1 x 109 Celdas