Learn More
STEMCELL Technologies EasySep™ Human Pan-DC Pre-Enrichment Kit
Código de producto. 17157911
Encompass
Change view
Click to view available options
Cantidad:
1 x 109 celdas
Tamaño de la unidad:
1 set
17157911 1 x 109 celdas 1 set
1 options
Código de producto. 17157911
Marca: STEMCELL Technologies 19251
Immunomagnetic isolation of untouched human pan dendritic cells (DC)
  • The EasySep™ Human Pan-DC Pre-Enrichment Kit is designed to pre-enrich all dendritic cells (DCs) (including myeloid and plasmacytoid DCs) from fresh or previously frozen peripheral blood mononuclear cells by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-pan DCs and dextran-coated magnetic particles
  • The labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
Especificaciones
Tipo de célula Dendritic Cells
Para utilizar con (aplicación) Cell Isolation
Cantidad 1 x 109 celdas