Learn More
STEMCELL Technologies EasySep™ Human EpCAM Positive Selection Kit II
Código de producto. 17117761
Encompass
Change view
Click to view available options
:
1 x 109 cells
Código de producto.
17117761 1 x 109 cells
1 options
Código de producto. 17117761
Marca: STEMCELL Technologies 17846
Immunomagnetic positive selection of human EpCAM+ cells (e.g. mammary epithelial cells)
  • The EasySep™ Human EpCAM Positive Selection Kit II is designed to isolate EpCAM+ cells from fresh or previously frozen human mammary epithelial cell preparations by immunomagnetic positive selection
  • Desired cells are targeted with antibody complexes recognizing EpCAM and dextran-coated magnetic particles
  • The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The EpCAM+ fraction contains luminal cells, luminal-restricted, and bipotential progenitor cells
  • This product replaces the EasySep™ Human EpCAM Positive Selection Kit (Catalog #18356)
Especificaciones
Tipo de célula Mammary Cells
Para utilizar con (aplicación) Cell Isolation
Cantidad 1 x 109 cells