Learn More
STEMCELL Technologies EasySep™ Human CD45 Depletion Kit II
Click to view available options
Cantidad:
2 x 109 celdas
Tamaño de la unidad:
1 set
Descripción
- The EasySep™ Human CD45 Depletion Kit II is designed to deplete CD45+ cells from fresh or previously frozen human peripheral blood mononuclear cells
- Unwanted cells are targeted with antibody complexes recognizing CD45 and magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Unwanted cells remain in the tube while desired cells are poured off
- The CD45 antigen is expressed on all hematopoietic cells except erythrocytes and platelets
- This product replaces the EasySep™ Human CD45 Depletion Kit (Catalog #18259) for even faster cell isolations
Especificaciones
Especificaciones
| Tipo de célula | Cancer Cells and Cell Lines |
| Para utilizar con (aplicación) | Cell Isolation |
| Cantidad | 2 x 109 celdas |