missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAAT1/GLAST-1/SLC1A3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 18 publications
Marca: Novus Biologicals NB100-1869
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
EAAT1/GLAST-1/SLC1A3 Polyclonal specifically detects EAAT1/GLAST-1/SLC1A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Especificaciones
| EAAT1/GLAST-1/SLC1A3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLAST, GLASTGLAST1, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3 | |
| Rabbit | |
| 60 kDa | |
| 0.1 mL | |
| Cellular Markers, Cognition and Behavior | |
| 6507 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 2 μg/mL, Flow Cytometry 1:200-1:500, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen 1:10-1:500, Flow (Intracellular) 1:500 | |
| P24942 | |
| SLC1A3 | |
| A synthetic peptide made to a C-terminal portion of the rat SLC1A3 protein (between residues 500-542) [Uniprot: P24942] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, rat and mouse. Based upon 100% immunogen sequence similarity, this antibody is predicted to cross-react with Bovine also. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido