Learn More
Dynein light chain 2 cytoplasmic Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Novus Biologicals NBP2-92136-0.1ml
 Ver más versiones de este producto
Código de producto. 18667481
  • 463.00€ / 0.10 ml
Descripción
Dynein light chain 2 cytoplasmic Polyclonal antibody specifically detects Dynein light chain 2 cytoplasmic in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Dynein light chain 2 cytoplasmic
Polyclonal
Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
DLC2, DLC8b, DNCL1B, dynein light chain 2, cytoplasmic, Dynein light chain LC8-type 2,8 kDa dynein light chain b, dynein, light chain, LC8-type 2, MGC17810, radial spoke 22 homolog, RSPH22
Recombinant fusion protein containing a sequence corresponding to amino acids 1-89 of human DYNLL2 (NP_542408.1). MSDRKAVIKNADMSEDMQQDAVDCATQAMEKYNIEKDIAAYIKKEFDKKYNPTWHCIVGRNFGSYVTHETKHFIYFYLGQVAILLFKSG
0.1 mL
Autophagy, Cell Biology, Cytoskeleton Markers, Signal Transduction
140735
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunofluorescence
Unconjugated
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified