missing translation for 'onlineSavingsMsg'
Learn More

DYNC1LI1 Monoclonal Antibody (OTI1H4), TrueMAB™, OriGene

Product Code. 17574441
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Unit Size:
100 microlitros
30 microlitros
Product Code. Cantidad unitSize
17574441 30 μL 30 microlitros
17554441 100 μL 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 17574441

Brand: Origene Technologies TA501511S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo requiere una actualización de precios antes de ser solicitado.

Solicite el precio de este artículo aquí.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DYNC1LI1 Monoclonal antibody specifically detects DYNC1LI1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antígeno DYNC1LI1
Aplicaciones Flow Cytometry, Immunocytochemistry, Western Blot
Clasificación Monoclonal
Clon OTI1H4
Concentración 0.4 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica DYNC1LI1
N.º de referencia del gen Q9Y6G9
Alias de gen 1110053F02Rik; Cytoplasmic dynein 1 light intermediate chain 1; DLC-A; DNCLI1; Dnclic1; Dync1li1; dynein cytoplasmic 1 light intermediate chain 1; dynein light chain A; Dynein light intermediate chain 1, cytosolic; dynein, cytoplasmic, light intermediate chain 1; dynein, cytoplasmic, light intermediate polypeptide 1; LIC1; LIC-1
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human DYNC1LI1 produced in HEK293T cell.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 51143
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.