Learn More
DYNC1LI1 Monoclonal Antibody (OTI1H4), TrueMAB™, OriGene
Código de producto. 17554441
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Tamaño de la unidad:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17554441 100 μL 100 microlitros
17574441 30 μL 30 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17554441 Proveedor Origene Technologies N.º de proveedor TA501511
Este artículo requiere una actualización de precios antes de ser solicitado.
Solicite el precio de este artículo aquí.
Mouse Monoclonal Antibody
Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules. May play a role in binding dynein to membranous organelles or chromosomes. Probably involved in the microtubule-dependent transport of pericentrin. Is required for progress throuh the spindle assembly checkpoint. The phosphorylated form appears to be involved in the selective removal of MAD1L1 and MAD1L2 but not BUB1B from kinetochores.
Especificaciones
Antígeno DYNC1LI1
Aplicaciones Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Clasificación Monoclonal
Clon OTI1H4
Concentración 0.4 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica DYNC1LI1
N.º de referencia del gen Q9Y6G9
Alias de gen DLC-A, DNCLI1, LIC1
Símbolos de los genes DYNC1LI1
Especie del huésped Mouse
Inmunógeno Full length human recombit protein of human DYNC1LI1 produced in HEK293T cell.
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 51143
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b