missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ CD14

Código de producto. p-200105791
Click to view available options
Cantidad:
2 mL
5 mL
Tamaño de la unidad:
2 ml
cada uno
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30276624

Marca: Invitrogen™ 11150D

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Dynabeads CD14 are superparamagnetic beads coupled with an anti-human CD14 antibody that enable easy isolation or depletion of human CD14+ cells directly from any sample, including whole blood, bone marrow, buffy coat, PBMC, and tissue digests.

Dynabeads CD14 are superparamagnetic beads coupled with an anti-human CD14 antibody that enable easy isolation or depletion of human CD14+ cells directly from any sample, including whole blood, bone marrow, buffy coat, peripheral blood mononuclear cells (PBMCs), and tissue digests. Advantages of Dynabeads CD14:

  • Fast and efficient isolation of CD14+ cells from any sample—no columns required
  • High purity, yield, and viability of positively isolated CD14+ cells for molecular downstream assays
  • Fast and efficient depletion of CD14+ cells from any sample

About Dynabeads CD14

Dynabeads CD14 are uniform, superparamagnetic beads (4.5 μm diameter) coated with a primary monoclonal antibody specific for the CD14 membrane antigen mainly expressed mainly on human monocytes. Because of the bead size, Dynabeads CD14 can easily and efficiently isolate or deplete cells from viscous samples such as whole blood and bone marrow in about 30 minutes. Dynabeads CD14 will also afford excellent recovery of high-purity, viable cells for use in downstream molecular studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, so if you want to isolate CD14+ cells for cell-based applications, or you need to check your samples with flow cytometry, check out our total human monocyte product offerings.

Magnetic bead to based separation offers easy handling

Dynabeads CD14 is added to the sample under continuous mixing to optimize the binding of the Dynabeads to the target cells. By placing the sample on a magnet, the bead-bound target cells are separated from the rest of the sample in just 1 to 2 minutes. For depletion, remove the supernatant to a new tube for further studies and discard the bead-bound cells. For positive isolation for molecular studies, remove the supernatant and wash the bead-bound cells 2 to 3 times in buffer to get optimal purity. The cells can be lysed while still attached to the beads and the supernatant transferred to a new tube for downstream molecular analysis. Starting samples can be whole blood, buffy coat, bone marrow, PBMC, or tissue digests.

Automation-ready for use with KingFisher purification systems

The product is ideal for high-throughput workflows using KingFisher systems. A user guide and scripts for KingFisher Flex and Apex workflows are available for download on this page.

Learn more about Dynabeads products

TRUSTED_SUSTAINABILITY

Especificaciones

Tipo de célula Monocytes
Contenido y almacenamiento This Product Contains: 2 mL Dynabeads™ CD14 coated with anti-CD14 monoclonal antibody.
Storage Conditions: 2°C to 8°C. Do not freeze.
Color Brown
Para utilizar con (aplicación) Cell Depletion, Positive Isolation for Molecular Applications
Material Magnetic Polystyrene
Tipo de producto Antibody Coated Bead
Compatibilidad de alto rendimiento Yes
N.° de celdas Processes ∼8x 10^8 cells total
Viabilidad de la salida >95%
Línea de productos Dynabeads
Grado de pureza o calidad Research Grade
Cantidad 2 mL
Tipo de muestra PBMC, Blood
Condiciones de envío Room Temperature
N.º de celda del material de partida 1 x 10^7 PBMCs per isolation
Especies diana Human
Diámetro (métrico) 4.5 μm
Mostrar más Mostrar menos

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado