Learn More
GA International DYMO-Compatible Cryogenic Labels for Frozen Surfaces
Descripción
- Quick labeling of frozen vials and tubes (no thawing required)
- Withstand exposure to solvents, water baths, and condensation
- Compatible with the 450 series and 4XL DYMO™ LabelWriters
- Convenient to print, no need for ribbons or other ink source
Especificaciones
Especificaciones
| Tipo de adhesivo | Permanent |
| Color | White |
| Acabado | Medium Gloss |
| Formato | Rollo |
| Material | Thermoplastic Film |
| Print Technology | Direct Thermal |
| Tipo de producto | Label |
| Esterilidad | No estéril |
| Etiquetas por fila | 1 Label |
| Cantidad | 2000/Pk. |
| Mostrar más |